missing translation for 'onlineSavingsMsg'
Learn More

TIGIT Monoclonal Antibody (GIGD7), Super Bright™ 702, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 67-9501-82

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933784

  • € 333.00 / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Rat Monoclonal Antibody

Binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T cell activation by promoting the generation of mature immunoregulatory dendritic cells.
Specifications

Specifications

TIGIT
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
P86176
TIGIT
mTIGIT-Fc treated with 4% PFA
100 μg
Primary
Mouse
Antibody
IgG2a κ
Flow Cytometry
GIGD7
Super Bright 702
TIGIT
ENSMUSG00000071552; T cell immunoreceptor with Ig and ITIM domains; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; Washington University cell adhesion molecule; WUCAM
Rat
Affinity chromatography
RUO
100043314
4° C, store in dark, DO NOT FREEZE!
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers