missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIGIT Recombinant Rabbit Monoclonal Antibody (BLR047F)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544290
This item is not returnable.
View return policy
Description
TIGIT Recombinant Monoclonal Antibody for Western Blot, IHC (P)
TIGIT is a 26 kDa protein that belongs to the poliovirus receptor (PVR) family. It is expressed on NK cells, regulatory T cells, follicular T helper cells, memory CD4+ T cells, and CD8+ T cells, but not on B cells or naive CD4+ T cells. However, TIGIT can be upregulated on naive CD4+ T cells upon activation. TIGIT interacts with members of the PVR and PVR-like families, including CD155. This interaction mediates the engagement of NK and T cells with antigen-presenting cells, fibroblasts, and endothelial cells expressing PVR and PVR-like proteins. Binding of TIGIT to PVR leads to increased secretion of IL-10 and decreased secretion of IL-12B, promoting the generation of mature immunoregulatory dendritic cells and suppressing T cell activation. Through these interactions, TIGIT plays a significant role in modulating immune responses and maintaining immune homeostasis.
Specifications
| TIGIT | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| BBS with 0.09% sodium azide; pH 8.2 | |
| Q495A1 | |
| TIGIT | |
| A peptide representing a region between residue 194 and the C-terminus (residue 244) of human T-cell immunoreceptor with Ig and ITIM domains precursor (TIGIT). | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| BLR047F | |
| Unconjugated | |
| TIGIT | |
| ENSMUSG00000071552; T cell immunoreceptor with Ig and ITIM domains; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; Washington University cell adhesion molecule; WUCAM | |
| Rabbit | |
| Purified | |
| RUO | |
| 201633 | |
| 2-8°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction