missing translation for 'onlineSavingsMsg'
Learn More

Tiglyl Glycine-13C2,15N, TRC

Product Code. 30006133
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006133 1 mg 1mg
30005050 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006133 Supplier Toronto Research Chemicals Supplier No. TRCT4401021MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula 13C2 C5 H11 15N O3
Molecular Weight (g/mol) 160.146
Synonym N-[(2E)-2-Methyl-1-oxo-2-buten-1-yl]glycine-13C2,15N, N-Tigloylglycine-13C2,15N, N-Tiglylglycine-13C2,15N;
IUPAC Name 2-[[(E)-2-methylbut-2-enoyl]amino]acetic acid
SMILES C\C=C(/C)\C(=O)[15NH][13CH2][13C](=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C7H11NO3/c1-3-5(2)7(11)8-4-6(9)10/h3H,4H2,1-2H3,(H,8,11)(H,9,10)/b5-3+/i4+1,6+1,8+1
Formula Weight 160.0776
Percent Purity >95%
Chemical Name or Material Tiglyl Glycine-13C2,15N
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.