missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tilorone dihydrochloride, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Tilorone dihydrochloride is the first recognized synthetic, small molecular weight compound that is an orally active interferon inducer, used as an antiviral drug.
Specifications
Specifications
| Chemical Name or Material | Tilorone dihydrochloride |
| Physical Form | Solid |
| CAS | 27591-69-1 |
| Color | Orange |
| Quantity | 500 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C25H36Cl2N2O3 |
| For Use With (Application) | COVID-19-anti-virus |
| Solubility Information | H2O : 100 mg/mL (206.84 mM; Need ultrasonic) ∣DMSO : 12.5 mg/mL (25.85 mM; Need ultrasonic) |
| SMILES | O=C1C2=C(C3=C1C=C(OCCN(CC)CC)C=C3)C=CC(OCCN(CC)CC)=C2.[H]Cl.[H]Cl |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction