Learn More
TIM-1/KIM-1/HAVCR Antibody (2389F), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals FAB17504B
Product Code. 18614843
  • ONLINE EXCLUSIVE PRICE
    € 647.00
    € 611.10 / 0.10mL
    Save € 35.90
    5% Off
Description
TIM-1/KIM-1/HAVCR Monoclonal antibody specifically detects TIM-1/KIM-1/HAVCR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TIM-1/KIM-1/HAVCR
Monoclonal
Biotin
PBS
Rabbit
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, Immunohistochemistry, CyTOF
2389F
Flow Cytometry, Immunohistochemistry, CyTOF-ready
HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
Mouse myeloma cell line NS0-derived human TIM-1/KIM-1/HAVCR, Pro21-Thr288, Accession # AAC39862
0.1 mL
CD Markers, Cellular Signaling, Immunology
26762
Store at 4°C in the dark.