missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-1/KIM-1/HAVCR Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56421DL594
This item is not returnable.
View return policy
Description
TIM-1/KIM-1/HAVCR Polyclonal antibody specifically detects TIM-1/KIM-1/HAVCR in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| TIM-1/KIM-1/HAVCR | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| DyLight 594 | |
| HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
| A synthetic peptide (SPLYSYTTDGNDTVT) of human TIM-1 protein was used as immunogen for this antibody. This sequence corresponds to amino acids (aa) 248-262 of NP_036338 and aa 84-98 of EAW61615.1. Isoforms of TIM-1 have been described. For example, NP_036338 is 364 aa and EAW61615.1 is 194 aa. EAW61615.1 is referred to as isoform CRA_a. | |
| 0.1 mL | |
| CD Markers, Cellular Signaling, Immunology | |
| 26762 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction