Learn More
TIM-1/KIM-1/HAVCR Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-56421FR
Product Code. 18547161
  • ONLINE EXCLUSIVE PRICE
    € 570.00
    € 538.65 / 0.10mL
    Save € 31.35
    5% Off
Description
TIM-1/KIM-1/HAVCR Polyclonal antibody specifically detects TIM-1/KIM-1/HAVCR in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TIM-1/KIM-1/HAVCR
Polyclonal
PBS
Rabbit
Protein G purified
RUO
Primary
Human, Mouse
Purified
Western Blot, Immunocytochemistry/Immunofluorescence
DyLight 680
HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
A synthetic peptide (SPLYSYTTDGNDTVT) of human TIM-1 protein was used as immunogen for this antibody. This sequence corresponds to amino acids (aa) 248-262 of NP_036338 and aa 84-98 of EAW61615.1. Isoforms of TIM-1 have been described. For example, NP_036338 is 364 aa and EAW61615.1 is 194 aa. EAW61615.1 is referred to as isoform CRA_a.
0.1 mL
CD Markers, Cellular Signaling, Immunology
26762
Store at 4°C in the dark.