missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-1/KIM-1/HAVCR Antibody (KIM70), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32775AF647
This item is not returnable.
View return policy
Description
TIM-1/KIM-1/HAVCR Monoclonal specifically detects TIM-1/KIM-1/HAVCR in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.
Specifications
| TIM-1/KIM-1/HAVCR | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HAVCR1 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with human recombinant KIM-1 ectodomain expressed in baculovirus system. | |
| 0.2 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| KIM70 | |
| Western Blot, ELISA, Sandwich ELISA | |
| HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 26762 | |
| Store at 4C in the dark. | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering