missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-1/KIM-1/HAVCR Rat anti-Mouse, Clone: 222414, R&D Systems™

Rat Monoclonal Antibody has been used in 11 publications
Brand: R&D Systems MAB1817-100
This item is not returnable.
View return policy
Description
TIM-1/KIM-1/HAVCR Monoclonal specifically detects TIM-1/KIM-1/HAVCR in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| TIM-1/KIM-1/HAVCR | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| NP_001160104 | |
| HAVCR1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TIM-1/KIM-1/HAVCR Tyr22-Thr212 Accession # NP_001160104 | |
| 100 μg | |
| Primary | |
| Detects mouse TIM-1/KIM-1/HAVCR in ELISAs and Western blots. This antibody does not cross-react with recombinant human (rh) TIM-1, rmTIM-2, or rmTIM-3. | |
| Mouse | |
| Purified |
| Western Blot, ELISA | |
| 222414 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 26762 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction