missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIM-1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA546914
This item is not returnable.
View return policy
Description
In sandwich ELISAs, less than 0.1% cross-reactivity with recombinant mouse (rm) TIM-1, recombinant rat TIM-1, and recombinant human (rh) TIM-4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CD365 also known as T-cell immunoglobulin and mucin domain 1 (Tim-1) is a type 1 surface glycoprotein that consists of extracellular N-terminal immunoglobulin V-like domain and a mucin domain, and a short cytoplasmic tail. The endogenous ligand for CD365 is Tim-4. In addition, CD365 is known to be a cellular receptor or co-receptor for a number of viruses including hepatitis A and C viruses, Ebola virus, Marburg virus, Dengue virus, Zika virus, West Nile virus, and Japanese encephalitis virus. The direct binding of the viruses by CD365 occurs via phosphatildyl-serine exposed on viral envelopes. Polymorphisms within human CD365 gene (HAVCR1) are associated with susceptibility to infection and the disease severity. CD365 has also been implicated in the T cell co-stimulation. CD365 is primarily expressed in epithelial cells. It is also highly expressed in A549, 769-P and Caco-2 and cell lines. CD365 exists also as a soluble protein generated by metalloproteinase mediated proteolytic cleavage of the surface protein.
Specifications
| TIM-1 | |
| Polyclonal | |
| Unconjugated | |
| Havcr1 | |
| AI503787; CD365; HAVCR; HAVCR1; HAVcr-1; Hepatitis A virus cellular receptor 1; hepatitis A virus cellular receptor 1 homolog; Kidney injury molecule 1; Kim1; KIM-1; T cell immunoglobin domain and mucin domain protein 1; t cell immunoglobulin and mucin domain-containing protein 1; T cell membrane protein 1; T-cell immunoglobulin and mucin domain containing 1; T-cell immunoglobulin and mucin domain-containing protein 1; T-cell immunoglobulin mucin family member 1; T-cell immunoglobulin mucin receptor 1; T-cell membrane protein 1; TIM; TIM1; TIM-1; TIMD1; TIMD-1 | |
| Goat | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26762 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q96D42 | |
| Havcr1 | |
| Mouse myeloma cell line NS0-derived recombinant human TIM-1/KIM-1/HAVCR Ser21-Thr288. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction