missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-2 Antibody (RM0134-5B23), DyLight 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22510DL594
This item is not returnable.
View return policy
Description
TIM-2 Monoclonal antibody specifically detects TIM-2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TIM-2 | |
| Monoclonal | |
| DyLight 594 | |
| T Cell immunoglobulin and mucin domain protein 2, Tim2 | |
| Mouse recombinant protein of TIM-2 | |
| 0.1 mL | |
| Cellular Signaling, Immunology | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0134-5B23 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction