missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-3 Antibody (024), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89462CL1
This item is not returnable.
View return policy
Description
TIM-3 Monoclonal antibody specifically detects TIM-3 in Human samples. It is validated for ELISA,Sandwich ELISA Capture
Specifications
| TIM-3 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human TIM-3 (Accession#: AAL65157.1; Met 1-Arg 200). | |
| 0.1 mL | |
| Cancer, Immunology, Innate Immunity, Neuroscience | |
| 84868 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Sandwich ELISA Capture | |
| 024 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction