missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-3 Antibody (TIM3/6863R), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24217UV
This item is not returnable.
View return policy
Description
TIM-3 Monoclonal antibody specifically detects TIM-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TIM-3 | |
| Monoclonal | |
| DyLight 350 | |
| CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
| Recombinant fragment of human TIM-3 protein (around aa1-200) (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Immunology, Innate Immunity, Neuroscience | |
| 84868 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| TIM3/6863R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction