missing translation for 'onlineSavingsMsg'
Learn More

TIM-3 (CD366) Mouse anti-Human, PE-CF594, Clone: 7D3, BD Horizon™

Mouse Monoclonal Antibody

€ 131.04 - € 351.92

Specifications

Antigen TIM-3 (CD366)
Clone 7D3
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE-CF594
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15814399
missing translation for 'viewDocuments'
BD Biosciences
565561
25 Tests
€ 131.04
Each
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
15804399
missing translation for 'viewDocuments'
BD Biosciences
565560
100 Tests
ONLINE EXCLUSIVE PRICE
€ 352.00
€ 351.92 / Each
Save € 0.08
0.02% Off
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.

TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.
Specifications

Specifications

TIM-3 (CD366)
Flow Cytometry
PE-CF594
Mouse
Cell Biology
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human TIM-3
Primary
Vol. Per test: 5μL
7D3
Monoclonal
Purified
RUO
Human
CD366; HAVCR2; TIM3; T cell immunoglobulin mucin-3; TIMD-3; KIM-3
IgG1 κ
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Videos
SDS
Documents

Documents

Certificates

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.