missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIM-4 Monoclonal Antibody (2A1H5)

Description
TIM-4 Monoclonal Antibody for Western Blot, ICC/IF, Flow, ELISA
The T cell immunoglobulin and mucin domain containing protein (TIM) family encodes cell surface receptors that are involved in the regulation of T helper (Th) -1 and -2 cell-mediated immunity. TIM-4, which is preferentially expressed on macrophages and dendritic cells, is the natural ligand of TIM-1, and this binding leads to T-cell expansion and cytokine production. Unlike other members of the TIM family, TIM-4 lacks a putative tyrosine phosphorylation signal sequence in its intracellular domain. The TIM-4 gene maps to a locus associated with predisposition to asthma in both mice and humans and with its connection to TIM-1-triggered Th2 responsiveness, may be considered as a candidate disease/predisposition gene for asthma.
Specifications
Specifications
| Antigen | TIM-4 |
| Applications | ELISA, Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2A1H5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | TIMD4 |
| Gene Accession No. | Q96H15 |
| Gene Alias | B430010N18Rik; RGD1564516; smuckler; soluble TIM 4; Spleen, mucin-containing, knockout of lymphotoxin protein; sTIM 4; T cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain-containing protein 4; T-cell immunoglobulin mucin receptor 4; T-cell membrane protein 4; Tim4; TIM-4; TIMD4; TIMD-4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?