missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-4, Rat anti-Mouse, Clone: 5G3, Novus Biologicals™
Rat Monoclonal Antibody
€ 281.00 - € 595.00
Specifications
| Antigen | TIM-4 |
|---|---|
| Clone | 5G3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Block/Neutralize |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Blocking Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18092816
|
Novus Biologicals
NBP2-81122-0.025MG |
0.025 mg |
€ 281.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18362350
|
Novus Biologicals
NBP2-81122-0.2MG |
0.2 mg |
€ 595.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIM-4 Monoclonal specifically detects TIM-4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Block/Neutralize.Specifications
| TIM-4 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Blocking Assay | |
| Unconjugated | |
| Rat | |
| Mouse | |
| FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD-4 | |
| TIMD4 | |
| IgG1 κ | |
| Protein A purified | |
| This antibody is specific for murine TIM-4, which unlike other TIM family members, is not expressed on T lymphocytes but rather found on antigen presenting cells such as lymphoid and myeloid DC's and macrophages. TIM-4 has been shown to interact with TIM-1 thereby suggesting a role of TIM-4 in the balance of Th1/Th2 regulation. Additionally TIM-4 has been shown to be a receptor for phosphatidylserine and may play a role in engulfment of apoptotic cells. |
| 5G3 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Block/Neutralize | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 91937 | |
| This antibody was raised by immunising female Lewis rats (Harlan Sprague-Dawley) with Tim-4:Ig fusion protein in complete Freund's adjuvant. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title