Learn More
TIMELESS Mouse anti-Human, Unconjugated, Clone: 1B6G3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67022-1-IG-150UL
Description
The TIMELESS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TIMELESS. This antibody recognizes human, mouse, rat antigen. The TIMELESS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TIMELESS | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, ELISA | |
| 1B6G3 | |
| Unconjugated | |
| 150 μL | |
| 8914 | |
| IgG1 |