Learn More
Amarell™ Multi-purpose Electronic Standard Thermometer
Description
- Practical and easy readability
- All-round thermometer for the fast and exact temperature indication
- Hold feature
- Automatic power-off after 10 min.
- Extremely fast responding semi-conductor stainless-steel probe
Specifications
Specifications
| Dimensions (L x W x H) | 22 x 55 mm |
| Disposable | Reusable |
| Length (Metric) Probe | 125 mm |
| Measurement Time | T90 = 4 sec. |
| Probe Type | Piercing |
| Resolution | 0.1°C |
| Product Type | Multi-electronic Standard Thermometer |
| Max. Temperature (Metric) | 200°C |
| Min. Temperature (Metric) | -50°C |
| Temperature Range (Metric) | -50°C to +200°C |
| Show More |