missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMM17B Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98507AF405
This item is not returnable.
View return policy
Description
TIMM17B Polyclonal specifically detects TIMM17B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| TIMM17B | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| inner mitochondrial membrane preprotein translocase, JM3, mitochondrial import inner membrane translocase subunit Tim17-B, TIM17BDXS9822, translocase of inner mitochondrial membrane 17 homolog B (yeast) | |
| Produced in rabbits immunized with E. coli-derived Human TIMM17B fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 10245 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction