missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TIMM17B Polyclonal Antibody

Product Code. 30019792
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30019792 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30019792 Supplier Invitrogen™ Supplier No. PA5148312

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TIMM17B Polyclonal Antibody for ICC/IF

This gene encodes a multipass transmembrane protein that forms an integral component of the mitochondrial translocase TIM23 complex. This complex facilitates the transport of mitochondrial proteins from the cytosol across the mitochondrial inner membrane and into the mitochondrion. There is a pseudogene for this gene on chromosome 12. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM17B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene Timm17b
Gene Accession No. O60830
Gene Alias 17kDa; DXS9822; inner mitochondrial membrane preprotein translocase; JM3; Mitochondrial import inner membrane translocase subunit Tim17-B; mTim17b; RGD1561744; Sfc3; TIM17B; TIMM17B; translocase of inner mitochondrial membrane 17 homolog B (yeast); translocase of inner mitochondrial membrane 17b; translocator of inner mitochondrial membrane 17 kDa, b; translocator of inner mitochondrial membrane 17b; translocator of inner mitochondrial membrane b
Gene Symbols Timm17b
Host Species Rabbit
Immunogen E. coli-derived Human TIMM17B fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10245
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.