missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIMM22 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104322
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TIM22.
TIM22 is a translocase localized in mitochondrial inner membrane. It is implicated in import of nuclear-encoded mitochondrial preproteins to the mitochondria. TIM22 is the essential core of protein insertion complex (TIM22 complex) that consists of TIM22, TIM54, TIM18, and TIM12.
Specifications
| TIMM22 | |
| Polyclonal | |
| Unconjugated | |
| Timm22 | |
| mitochondrial import inner membrane translocase subunit Tim22; preprotein translocase; putative membrane protein; Testis-expressed protein 4; testis-expressed sequence 4; TEX4; TIM22; TIMM22; translocase of inner mitochondrial membrane 22; translocase of inner mitochondrial membrane 22 homolog; translocase of inner mitochondrial membrane 22 homolog (yeast) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 29928, 56322, 79463 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9CQ85, Q9JKW1, Q9Y584 | |
| Timm22 | |
| A synthesized peptide derived from human TIMM22(Accession Q9Y584), corresponding to amino acid residues D95-S145. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction