missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMM23 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 204.00 - € 470.00
Specifications
| Antigen | TIMM23 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18677701
|
Novus Biologicals
NBP2-93133-0.02ml |
0.02 mL |
€ 204.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690691
|
Novus Biologicals
NBP2-93133-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIMM23 Polyclonal antibody specifically detects TIMM23 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| TIMM23 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| bA592B15.7, MGC22767, PRO1197, RP11-592B15.7, TIM23 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 7-72 of human TIMM23 (NP_006318.1). SGNKTTGGLAGFFGAGGAGYSHADLAGVPLTGMNPLSPYLNVDPRYLVQDTDEFILPTGANKTRGR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 100287932 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title