missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIMM23 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5141027
This item is not returnable.
View return policy
Description
TIMM23 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Essential component of the TIM23 complex, a complex that mediates the translocation of transit peptide-containing proteins across the mitochondrial inner membrane.
Specifications
| TIMM23 | |
| Polyclonal | |
| Unconjugated | |
| Timm23 | |
| mitochondrial import inner membrane translocase subunit Tim23; TIM23; TIMM23; translocase of inner mitochondrial membrane 23; translocase of inner mitochondrial membrane 23 homolog; translocase of inner mitochondrial membrane 23 homolog (yeast); translocase of the inner mitochondrial membrane | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100287932, 53600 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.21 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O14925, Q9WTQ8 | |
| Timm23 | |
| Synthetic peptide within human Tim23 aa 30-80. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction