missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TIMM44 Recombinant Rabbit Monoclonal Antibody (8J1F10)

Product Code. 30132265
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132265 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30132265 Supplier Invitrogen™ Supplier No. MA555574

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen Sequence: WCRCPRRCLG SGIQFLSSHN LPHGSTYQMR RPGGELPLSK SYSSGNRKGF LSGLLDNVKQ ELAKNKEMKE SIKKFRDEAR RLEESDVLQE ARRKYKTIES ETVRTSEVLR KKLGELTGTV KESLHEVSKS DLGRKIKEGV EEAAKTAKQS AESVSKGGEK LGRTAAFRAL SQGVESVKKE IDDSVLGQTG PYRRPQRLRK RTEFAGDKFK EEKVFEPNEE ALGVVLHKDS K.

This gene encodes a peripheral membrane protein associated with the mitochondrial inner membrane translocase, which functions in the import of proteins across the mitochondrial inner membrane and into the mitochondrial matrix. The encoded protein mediates binding of mitochondrial heat shock protein 70 to the translocase of inner mitochondrial membrane 23 (TIM23) complex. Expression of this gene is upregulated in kidney in a mouse model of diabetes. A mutation in this gene is associated with familial oncocytic thyroid carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM44
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 8J1F10
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TIMM44
Gene Accession No. O35094, O43615
Gene Alias 0710005E20Rik; D8Ertd118e; Mimt44; Mitochondrial import inner membrane translocase subunit TIM44; Tim44; TIMM44; translocase of inner mitochondrial membrane 44; translocase of inner mitochondrial membrane 44 homolog; translocase of inner mitochondrial membrane 44 homolog (yeast); translocator of inner mitochondrial membrane 44; translocator of inner mitochondrial membrane 44; DNA segment, Chr 8, ERATO Doi 118, expressed
Gene Symbols TIMM44
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 10-240 of human TIM44.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10469, 29635
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.