missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIMM44 Recombinant Rabbit Monoclonal Antibody (JE65-55)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544741
This item is not returnable.
View return policy
Description
TIMM44 Recombinant Monoclonal Antibody for Western Blot
This gene encodes a peripheral membrane protein associated with the mitochondrial inner membrane translocase, which functions in the import of proteins across the mitochondrial inner membrane and into the mitochondrial matrix. The encoded protein mediates binding of mitochondrial heat shock protein 70 to the translocase of inner mitochondrial membrane 23 (TIM23) complex. Expression of this gene is upregulated in kidney in a mouse model of diabetes. A mutation in this gene is associated with familial oncocytic thyroid carcinoma.
Specifications
| TIMM44 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O35094, O43615 | |
| TIMM44 | |
| Recombinant protein within human TIM44 aa 1-280/452. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| JE65-55 | |
| Unconjugated | |
| TIMM44 | |
| 0710005E20Rik; D8Ertd118e; Mimt44; Mitochondrial import inner membrane translocase subunit TIM44; Tim44; TIMM44; translocase of inner mitochondrial membrane 44; translocase of inner mitochondrial membrane 44 homolog; translocase of inner mitochondrial membrane 44 homolog (yeast); translocator of inner mitochondrial membrane 44; translocator of inner mitochondrial membrane 44; DNA segment, Chr 8, ERATO Doi 118, expressed | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10469, 29635 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction