missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMM50 Monoclonal Antibody (OTI1E9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811603S
This item is not returnable.
View return policy
Description
TIMM50 Monoclonal antibody specifically detects TIMM50 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| TIMM50 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O08553, P47942, Q16555 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 12934, 1808, 25416 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI1E9 | |
| Unconjugated | |
| DPYSL2 | |
| 2810403L02Rik; AU015082; homolog of yeast Tim50; hypothetical protein LOC505489; LOC687295; LOW QUALITY PROTEIN: mitochondrial import inner membrane translocase subunit TIM50; MGC102733; mitochondrial import inner membrane translocase subunit TIM50; PRO1512; similar to translocase of inner mitochondrial membrane 50 homolog; TIM50; TIM50L; Tim50-like protein; timm50; timm50 protein; translocase of inner mitochondrial membrane 50; translocase of inner mitochondrial membrane 50 homolog; translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae); translocase of inner mitochondrial membrane 50-like protein; zgc:66357 | |
| Human recombinant protein fragment corresponding to amino acids 13-490 of human DPYSL2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction