missing translation for 'onlineSavingsMsg'
Learn More

TIMM8A Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18014959 Shop All Bio Techne Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
0.1 mL
missing translation for 'unitSize'
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18014959 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 18014959 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP298033AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TIMM8A Polyclonal specifically detects TIMM8A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM8A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DDP1deafness/dystonia peptide, DDPMGC12262, Deafness dystonia protein 1, DFN1, mitochondrial import inner membrane translocase subunit Tim8 A, MTSTIM8, TIM8A, translocase of inner mitochondrial membrane 8 (yeast) homolog A, translocase of inner mitochondrial membrane 8 homolog A (yeast), X-linked deafness dystonia protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TIMM8A fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1678
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.