missing translation for 'onlineSavingsMsg'
Learn More

Timonacic, TRC

Product Code. 30451023
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30451023

Brand: Toronto Research Chemicals TRCT4437351G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Timonacic
CAS 444-27-9
Molecular Formula C4 H7 N O2 S
Quantity 1 g
Synonym Thioproline, 4-Thiazolidinecarboxylic Acid, 1,3-Thiazolane-4-carboxylic Acid, 1,3-Thiazolidin-3-ium-4-carboxylate, 4-carboxythiazolidine, 1,3-Thiazolidine-4-carboxylic Acid
InChI Formula InChI=1S/C4H7NO2S/c6-4(7)3-1-8-2-5-3/h3,5H,1-2H2,(H,6,7)
SMILES OC(=O)C1CSCN1
IUPAC Name 1,3-thiazolidine-4-carboxylic acid
Molecular Weight (g/mol) 133.169
Formula Weight 133.02
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.