missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Timonacic, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Timonacic |
| CAS | 444-27-9 |
| Molecular Formula | C4 H7 N O2 S |
| Quantity | 500 mg |
| Synonym | Thioproline, 4-Thiazolidinecarboxylic Acid, 1,3-Thiazolane-4-carboxylic Acid, 1,3-Thiazolidin-3-ium-4-carboxylate, 4-carboxythiazolidine, 1,3-Thiazolidine-4-carboxylic Acid |
| InChI Formula | InChI=1S/C4H7NO2S/c6-4(7)3-1-8-2-5-3/h3,5H,1-2H2,(H,6,7) |
| SMILES | OC(=O)C1CSCN1 |
| IUPAC Name | 1,3-thiazolidine-4-carboxylic acid |
| Molecular Weight (g/mol) | 133.169 |
| Formula Weight | 133.02 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction