missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-1 Antibody (TIMP1/1944R), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08713DL594
This item is not returnable.
View return policy
Description
TIMP-1 Monoclonal antibody specifically detects TIMP-1 in Human samples. It is validated for ELISA
Specifications
| TIMP-1 | |
| Monoclonal | |
| DyLight 594 | |
| CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1 | |
| Recombinant human full-length TIMP-1 protein (Uniprot: P01033) | |
| 100 μg | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix | |
| 7076 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| TIMP1/1944R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction