missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TIMP-2 Antibody (3A4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30890-200ug
This item is not returnable.
View return policy
Description
TIMP-2 Monoclonal antibody specifically detects TIMP-2 in Chicken,Human,Mouse,Porcine,Rabbit,Zebrafish samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen)
Specifications
| TIMP-2 | |
| Monoclonal | |
| Unconjugated | |
| Phosphate buffered saline | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Chicken, Human, Mouse, Pig, Rabbit, Zebrafish | |
| Purified |
| ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 3A4 | |
| Western Blot : 1-2 μg/mL, ELISA, Immunohistochemistry-Paraffin : 1-5 μg/mL, Immunohistochemistry-Frozen : 1-5 μg/mL | |
| CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2 | |
| Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the human TIMP-2 protein and mouse myeloma Ag8563 cells. Sequence common in mouse, rabbit, chicken, zebrafish and pig. | |
| 200 μg | |
| Angiogenesis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers | |
| 7077 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction