missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-2 Antibody (TIMP2/2488R), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08721B
This item is not returnable.
View return policy
Description
TIMP-2 Monoclonal antibody specifically detects TIMP-2 in Human, Mouse, Rat, Bovine, Guinea Pig, Rabbit, Zebrafish samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TIMP-2 | |
| Monoclonal | |
| Biotin | |
| CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2 | |
| A synthetic peptide from the N-terminus of human TIMP-2 protein (aa56-66) (DSGNDIYGNPI) (Uniprot: P16035) | |
| 100 μg | |
| Angiogenesis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers | |
| 7077 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| TIMP2/2488R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Guinea Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction