missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-2 Antibody (TIMP2/8193R), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21033AF350
This item is not returnable.
View return policy
Description
TIMP-2 Monoclonal antibody specifically detects TIMP-2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TIMP-2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2 | |
| Recombinant fragment of human TIMP-2 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers | |
| 7077 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| TIMP2/8193R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction