Learn More
TIMP-3 Antibody (277128), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB9731H
Product Code. 18620305
  • € 608.00 / 0.10mL
Description
TIMP-3 Monoclonal antibody specifically detects TIMP-3 in Human samples. It is validated for Immunoprecipitation, Flow Cytometry, CyTOF-reported
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunoprecipitation, CyTOF
HRP
PBS
IgG2a
Monoclonal
Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-reported
Angiogenesis, Apoptosis, Cancer, Hypoxia, Metastasis, Vision