missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-3 Antibody (277128) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9731JF669
This item is not returnable.
View return policy
Description
TIMP-3 Monoclonal antibody specifically detects TIMP-3 in Human samples. It is validated for Immunoprecipitation,Flow Cytometry (Intracellular Staining),CyTOF-reported
Specifications
| TIMP-3 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| HSMRK222, K222, K222TA2, metalloproteinase inhibitor 3, MIG-5 protein, Protein MIG-5, pseudoinflammatory), SFD, TIMP metallopeptidase inhibitor 3, TIMP-3, tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, Tissue inhibitor of metalloproteinases 3 | |
| Mouse myeloma cell line NS0-derived recombinant human TIMP-3, Cys24-Pro211, Accession # P35625 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Immunoprecipitation, Flow Cytometry (Intracellular Staining), CyTOF | |
| 277128 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Cancer, Hypoxia, Metastasis, Vision | |
| 7078 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction