Learn More
TIMP2 Mouse anti-Human, Clone: OTI1B11, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF504044
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintece of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.Specifications
| TIMP2 | |
| Monoclonal | |
| Unconjugated | |
| TIMP2 | |
| CSC-21K, DDC8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7077 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1B11 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P16035 | |
| TIMP2 | |
| Full length human recombit protein of human TIMP2 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.