Learn More
enQuireBio™ Recombinant other Lypressin Protein
Product Code. 15940979
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
Product Code. 15940979
Brand: enQuireBio™ QP126085mg
Recombinant Protein
Specifications
Name Lypressin Protein
Quantity 5 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Other
Species E. coli
Protein Tag Untagged
Buffer The protein (1 mg/ml) was lyophilized with no additives.
Purity or Quality Grade Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.