Learn More
Invitrogen™ TIMP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595447
Description
Human TIMP4 shares 92.2% amino acid (aa) sequence identity with both mouse and rat TIMP4. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat heart tissue, mouse heart tissue, HELA whole cell. IHC: rat kidney tissue, human intestinal cancer tissue, human lung cancer tissue. ICC/IF: HepG2 cell. Flow: A549 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Tissue inhibitor of metalloproteinase 4 (TIMP4) is encoded by the gene TIMP4 located on chromosome 3. It belongs to the family of TIMP genes and is known to inhibit the activity of matrix metalloproteinases. Matrix metalloproteinases (MMP) are a group of proteinases that degrade the components of extracellular matrix. TIMP4 irreversibly binds to MMPs and inactivates them with the help of zinc as a cofactor. The netrin domain of TIMP4 is involved in platelet recruitment and aggregation. Other biological functions include response to cytokine and peptide hormone stimuli and involvement with the ovulation cycle and CNS development.
Specifications
| TIMP4 | |
| Polyclonal | |
| Unconjugated | |
| TIMP4 | |
| Metalloproteinase inhibitor 4; TIMP 4; TIMP metallopeptidase inhibitor 4; TIMP4; TIMP-4; tissue inhibitor of metalloproteinase 4; tissue inhibitor of metalloproteinases 4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 110595, 680130, 7079 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P81556, Q99727, Q9JHB3 | |
| TIMP4 | |
| E.coli-derived human TIMP4 recombinant protein (Position: N174-P224). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.