missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) 2,3-naphthalocyanine Dichloride, TRC

Product Code. 30452709
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452709 100 mg 100mg
30455625 250 mg 250mg
30455711 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30452709 Supplier Toronto Research Chemicals Supplier No. TRCT444088100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26857-61-4
Molecular Formula C21 H25 N7 O
Molecular Weight (g/mol) 391.469
Synonym (OC-6-12)-Dichloro[37H,39H-tetranaphtho[2,3-b:2',3'-g:2' ',3' '-l:2' '',3' ''-q]porphyrazinato(2-)-kN37,kN38,kN39,kN40]tin, 37H,39H-Tetranaphtho[2,3-b:2',3'-g:2' ',3' '-l:2' '',3' ''-q]porphyrazine, Tin complex, 37H,39H-Tetranaphtho[2,3-b:2',3'-g:2' ',3' '-l:2' '',3' ''-q]porphyrazine, Tin deriv., (OC-6-12)-Dichloro[37H,39H-tetranaphtho[2,3-b:2',3'-g:2' ',3' '-l:2' '',3' ''-q]porphyrazinato(2-)-N37,N38,N39,N40]-tin
IUPAC Name 1-[[4-(cyclopropylmethyl)-5-[2-(pyridin-4-ylamino)pyrimidin-4-yl]pyrimidin-2-yl]amino]-2-methylpropan-2-ol
SMILES CC(C)(O)CNc1ncc(c(CC2CC2)n1)c3ccnc(Nc4ccncc4)n3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H25N7O/c1-21(2,29)13-25-19-24-12-16(18(28-19)11-14-3-4-14)17-7-10-23-20(27-17)26-15-5-8-22-9-6-15/h5-10,12,14,29H,3-4,11,13H2,1-2H3,(H,24,25,28)(H,22,23,26,27)
Formula Weight 391.212
Chemical Name or Material Tin(IV) 2,3-naphthalocyanine Dichloride
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.