missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIN2 Antibody (SR1681), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | TIN2 |
|---|---|
| Clone | SR1681 |
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620675
|
Novus Biologicals
NBP3-22077-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648234
|
Novus Biologicals
NBP3-22077-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIN2 Monoclonal antibody specifically detects TIN2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| TIN2 | |
| Western Blot 1:500-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| 26277 | |
| IgG | |
| Affinity purified |
| SR1681 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| (TRF1)-interacting nuclear factor 2 variant 1, TERF1 (TRF1)-interacting nuclear factor 2, TERF1-interacting nuclear factor 2, TIN2TRF1-interacting nuclear protein 2 | |
| A synthesized peptide derived from human TIN2 (Uniprot #: Q9BSI4) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title