missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TINAG Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19747-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to TINAG.
Mediates adhesion of proximal tubule epithelial cells via integrins alpha3-beta1 and alphaV-beta3. This is a non catalytic peptidase C1 family protein.Specifications
| TINAG | |
| Polyclonal | |
| Unconjugated | |
| TINAG | |
| TIN AG, TINAG | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26944, 27283 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UJW2 | |
| Tinag | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction