missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tinoridine hydrochloride, MedChemExpress
Description
Tinoridine hydrochloride is a nonsteroidal anti-inflammatory drug and also has potent radical scavenger and antiperoxidative activity.
Specifications
Specifications
| Chemical Name or Material | Tinoridine hydrochloride |
| Physical Form | Solid |
| CAS | 25913-34-2 |
| Color | Yellow |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C17H21ClN2O2S |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | Y-3642 hydrochloride |
| Solubility Information | DMSO : 16.67 mg/mL (47.24 mM; Need ultrasonic) ∣H2O : 2.5 mg/mL (7.08 mM; ultrasonic and warming and heat to 60°C) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?