Learn More
Lovibond™ Tintometer™ CHECKIT™ Comparator Nitrate Test Tube
For use with Nitrate LR Checkit Comparator Kit.
Brand: Lovibond™ 366220
Additional Details : Weight : 0.01400kg
Description
For use with Nitrate LR Checkit Comparator Kit.
- The Lovibond™ Checkit™ comparator is a compact, lightweight colorimetric unit
- The D55 Checkit Comparator contains mirror optics which extend the cell path length to 55mm, allowing greater accuracy at lower concentrations
- Suitable for both mobile and static water analysis
- Available with a number of different color scales
- Provides the basis for a comprehensive easy to to to use analysis system
- Each Checkit™ Comparator Disc contains a continuous color scale for exact color matching between standard and sample
Specifications
Nitrate LR CHECKIT™ Comparator Kit | |
CHECKIT™ Comparator Nitrate Test Tube |