Learn More
Lovibond™ Tintometer™ Combipack Molybdate LR, Molybdate LR No. 1 / No. 2
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand:  Lovibond™ 517772
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 15331935
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Combi-Pack Molybdate LR
  • Molybdate LR No. 1and Molybdate LR No. 2
  • Available as two packs of 100 tablets or two packs of 250
Specifications
Specifications
Photometry Tablet Reagents
2 Packs of 250 Tablets
Combipack Molybdate LR, Molybdate LR No. 1 / No. 2
Test kits, colorimeters and spectrophotometers
250 x Molybdate LR No. 1; 250 x Molybdate LR No. 2 Tablets
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers