missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ Comparator 2000+ Disc 4/28, Hazen Scale, 50 to 500
Click to view available options
Range:
50 to 500 color Units
Unit Size:
Each
Description
- Color of water Disc 4/28
- Hazen Scale
- 50 to 500 color Units
Specifications
Specifications
| For Use With (Equipment) | Comparator 2000+ |
| Range | 50 to 500 color Units |
| Test Method | Hazen Scale |
| Type | Color of water Disc 4/28 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction