Learn More
Lovibond™ Tintometer™ Phosphate No. 2 HR Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Merk:  Lovibond™ 515821BT
Overige gegevens : Gewicht : 0.07500kg Transport : UN number : 2859 Chem class : 6.1 Pack group : II
 Bekijk meerdere types van dit product via de
Produktcode. 15331885
Dit item is uit de verkoop gehaald en is niet meer verkrijgbaar. Bekijk het product voor mogelijke alternatieven of neem contact op met ons technische ondersteuningsteam via 020-4877000 voor hulp.
Beschrijving
Beschrijving
  • New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
  • With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
  • Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
  • Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
  • ‘BT’ is added to the end of the code to identify the new style of packaging
  • Simple, time-saving and practical
  • There are no safety risks if the tablets are used in line with the instructions supplied
  • Safety data sheets are available for all reagents
Specificatie
Specificatie
250 Tablets
Test kits, colorimeters and spectrophotometers
Photometry Tablet Reagents
Phosphate No. 2 HR Tablets
Water Testing
Video's