Learn More
Lovibond™ Tintometer™ Thymol Blue Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
€ 34.90 - € 76.30
Specifications
For Use With (Application) | Photometry Tablet Reagents |
---|---|
For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
Type | Thymol Blue Tablets |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15331735
|
Lovibond™
511651BT |
250 Tablets |
€ 76.30
Pack of 250 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
15321735
|
Lovibond™
511650BT |
100 Tablets |
€ 34.90
Pack of 100 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
- Thymol Blue Tablets
- Available in packs of 100 and 250
Specifications
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
Thymol Blue Tablets |