Learn More
Sampling Systems™ Tip for Unit Dose Sleeve Sampler
Shop All Sampling Systems Products
Click to view available options
Length (Metric):
9.5 mm
11.5 mm
13.5 mm
14.5 mm
15.5 mm
17.5 mm
18.5 mm
19.5 mm
21.5 mm
22.5 mm
23.5 mm
25.5 mm
26.5 mm
27.5 mm
29.5 mm
30.5 mm
31.5 mm
34.5 mm
35.5 mm
38 mm
41.5 mm
45.5 mm
49 mm
52.5 mm
54 mm
Capacity (Metric):
0.1 mL
0.2 mL
0.3 mL
0.4 mL
0.5 mL
0.6 mL
0.7 mL
0.8 mL
0.9 mL
1.0 mL
1.1 mL
1.2 mL
1.3 mL
1.4 mL
1.5 mL
1.6 mL
1.7 mL
1.8 mL
1.9 mL
2 mL
2.1 mL
2.2 mL
2.3 mL
2.4 mL
2.5 mL
Diameter (Metric):
6 mm
8 mm
Unit Size:
Each
Description
- For sampling small volume powders and granules.
- Chemically resistant 316L stainless steel construction.
- Different sample sizes can be taken using the same sampler
- Tips for use with Type 1 or Type 2 Sampling Chambers
Specifications
Specifications
| Product Type | Unit Dose Sleeve Sampler Tip |
| Material | Stainless Steel (316L) |
| Capacity (Metric) | 1.7 mL |
| Diameter (Metric) | 8 mm |
| Length (Metric) | 25.5 mm |