missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PON1 Antibody (4G8D3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80919
This item is not returnable.
View return policy
Description
PON1 Monoclonal specifically detects PON1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, ICC/IF (Negative), CyTOF-ready.
Specifications
| PON1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 1:500 - 1:2000, Flow Cytometry : 1:200 - 1:400, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin, ICC/IF (Negative), CyTOF-ready | |
| A-esterase 1, Aromatic esterase 1, arylesterase B-type, EC 3.1.1.2, EC 3.1.1.81, EC 3.1.8.1, ESAesterase A, K-45, MVCD5, paraoxonase 1, PON 1, PONparaoxonase B-type, serum aryldiakylphosphatase, Serum aryldialkylphosphatase 1, serum paraoxonase/arylesterase 1 | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry, CyTOF | |
| 4G8D3 | |
| Unconjugated | |
| PBS | |
| PON1 | |
| Partial recombinant human PON1 (between residues 1-200) expressed in E. coli [Uniprot P27169] | |
| 0.1 mL | |
| Alzheimers Research, Cancer, Lipid and Metabolism, Neurodegeneration, Neuroscience | |
| 5444 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction