missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIP120A Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49918PE
This item is not returnable.
View return policy
Description
TIP120A Polyclonal antibody specifically detects TIP120A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| TIP120A | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| cullin-associated and neddylation-dissociated 1, Cullin-associated and neddylation-dissociated protein 1, cullin-associated NEDD8-dissociated protein 1, DKFZp434M1414, FLJ90441, KIAA0829FLJ10114, p120 CAND1, TBP interacting protein, TBP-interacting protein 120A, TBP-interacting protein of 120 kDa A, TIP120AFLJ38691, TIP120FLJ10929 | |
| The immunogen for this product maps to a region between residue 1180 and 1230 of human Cullin-Associated and Neddylation-Dissociated 1 using the numbering given in entry NP_060918.2 (GeneID 55832). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55832 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction